Learn More
Selleck Chemical LLC BRL-15572 Dihydrochloride S2677-5mg
SDP
Catalog No. 503110224
Encompass_Preferred
BRL-15572 is a 5-HT1D receptor antagonist with pKi of 7 9 also shows a considerable affinity at 5-HT1A and 5-HT2B receptors exhibiting 60-fold selectivity over 5-HT1B receptor
Catalog No. 50-311-0224 Supplier Selleck Chemical LLC Supplier No. S26775MG
BRL-15572 is a 5-HT1D receptor antagonist with pKi of 7 9 also shows a considerable affinity at 5-HT1A and 5-HT2B receptors exhibiting 60-fold selectivity over 5-HT1B receptor