Learn More
TARGETMOL CHEMICALS INC BRL-15572 dihydrochloride 25MG
SDP
Catalog No. 502877068
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. BRL-15572 dihydrochloride (BRL 15573 dihydrochloride) is a 5-HT1D receptor antagonist with pKi of 7.9, also shows a considerable affinity at 5-HT1A and 5-HT2B receptors, exhibiting 60-fold selectivity over 5-HT1B receptor. Purity 99.8%
Catalog No. 50-287-7068 Supplier TARGETMOL CHEMICALS INC Supplier No. T642325MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. BRL-15572 dihydrochloride (BRL 15573 dihydrochloride) is a 5-HT1D receptor antagonist with pKi of 7.9, also shows a considerable affinity at 5-HT1A and 5-HT2B receptors, exhibiting 60-fold selectivity over 5-HT1B receptor. Purity 99.8%