Learn More
Cayman Chemical BRL 37344sodIum salt 5mg
SDP
Catalog No. 502966227
Encompass_Preferred
A selective agonist of β3-adrenergic receptors with Ki values of 0.43, 9.17, and 37.9 µM for β3, β2, and β1, respectively
Catalog No. 50-296-6227 Supplier Cayman Chemical Supplier No. 186225 MG
A selective agonist of β3-adrenergic receptors with Ki values of 0.43, 9.17, and 37.9 µM for β3, β2, and β1, respectively