Learn More
Bellco Glass, Inc. Break Resistant Lip 3 Baffles Fernbach, 1800mL Side Baffles
SDP
Catalog No. 501945551
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 1800mlStopper Size: 10Case Quantity: 3
Catalog No. 50-194-5551 Supplier Bellco Glass, Inc. Supplier No. 255101800
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 1800mlStopper Size: 10Case Quantity: 3