Learn More
Bellco Glass, Inc. Break Resistant Lip 3 Baffles Fernbach, 2800mL Side Baffles
SDP
Catalog No. 501945552
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2800mlStopper Size: 13Case Quantity: 3 Dimensions: 210x225mm
Catalog No. 50-194-5552 Supplier Bellco Glass, Inc. Supplier No. 255102800
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 2800mlStopper Size: 13Case Quantity: 3 Dimensions: 210x225mm