Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, Plain 59mm ID Top, 3 Side Baffles, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 501214781
Encompass_Preferred
Flask, Fernbach, 2800mL, Plain 59mm ID Top, 3 Side Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on lower outside edge to help oxygen transfer to the culture medium.
Catalog No. 50-121-4781 Supplier Chemglass Life Sciences Supplier No. CLS202214
Flask, Fernbach, 2800mL, Plain 59mm ID Top, 3 Side Baffles, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on lower outside edge to help oxygen transfer to the culture medium.