Learn More
Medchemexpress LLC BRL-44408 maleate | 681806-46-2 | 99.0% | 10 MG
SDP
Catalog No. 5000341309
Encompass_Preferred
BRL-44408 maleate is a selective, blood-brain barrier-permeable α2A-adrenergic receptor antagonist with a Kᵢ value of 8.56 nM against human targets. It exhibits antidepressant and analgesic effects, and attenuates sepsis-induced acute lung injury by regulating neurotransmitter release or inhibiting signaling pathways. It can be used in studies related to acute respiratory distress syndrome, depression, and visceral pain.
  • Selective α2A-adrenergic receptor antagonist
  • Blood-brain barrier permeable
  • Exhibits antidepressant and analgesic effects
  • Attenuates sepsis-induced acute lung injury
  • Regulates neurotransmitter release and inhibits signaling pathways
  • Used in studies for acute respiratory distress syndrome, depression, and visceral pain
Catalog No. 50-003-41309 Supplier Medchemexpress LLC Supplier No. HY12716A10MG
Add to Cart
Add to Cart
BRL-44408 maleate is a selective, blood-brain barrier-permeable α2A-adrenergic receptor antagonist with a Kᵢ value of 8.56 nM against human targets. It exhibits antidepressant and analgesic effects, and attenuates sepsis-induced acute lung injury by regulating neurotransmitter release or inhibiting signaling pathways. It can be used in studies related to acute respiratory distress syndrome, depression, and visceral pain.
  • Selective α2A-adrenergic receptor antagonist
  • Blood-brain barrier permeable
  • Exhibits antidepressant and analgesic effects
  • Attenuates sepsis-induced acute lung injury
  • Regulates neurotransmitter release and inhibits signaling pathways
  • Used in studies for acute respiratory distress syndrome, depression, and visceral pain