Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BRL-44408 maleate | 681806-46-2 | MFCD02262218 | 99.0% | 331.37 g/mol | C17H21N3O4 | 5 MG
SDP

Catalog No. 5000264658
Encompass_Preferred

BRL-44408 maleate is a small-molecule α2A-adrenoceptor antagonist used in preclinical pharmacology research. It shows high affinity for α2A receptors (Ki ≈ 8.5 nM) and has demonstrated antidepressant and analgesic activity in animal studies.

  • Selective α2A-adrenoceptor antagonist with Ki ≈ 8.5 nM.
  • Demonstrates antidepressant and analgesic activity in preclinical models.
  • Solid, light brown to brown appearance.
  • Molecular weight 331.37 g/mol; formula C17H21N3O4.
  • Purity 99.0% (HPLC).
  • Soluble in DMSO at 30 mg/mL; may require ultrasonic and warming.
  • Store sealed, away from moisture at -20 °C; in solution, store at -80 °C for long-term stability.

Catalog No. 50-002-64658 Supplier Medchemexpress LLC Supplier No. HY12716A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BRL-44408 maleate is a small-molecule α2A-adrenoceptor antagonist used in preclinical pharmacology research. It shows high affinity for α2A receptors (Ki ≈ 8.5 nM) and has demonstrated antidepressant and analgesic activity in animal studies.

  • Selective α2A-adrenoceptor antagonist with Ki ≈ 8.5 nM.
  • Demonstrates antidepressant and analgesic activity in preclinical models.
  • Solid, light brown to brown appearance.
  • Molecular weight 331.37 g/mol; formula C17H21N3O4.
  • Purity 99.0% (HPLC).
  • Soluble in DMSO at 30 mg/mL; may require ultrasonic and warming.
  • Store sealed, away from moisture at -20 °C; in solution, store at -80 °C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.