Learn More
Abcam BRL 44408 maleate, alpha2A adrenoceptor antagonist, 10MG
Catalog No. 502707246
Encompass_Preferred
MW 331.37 g/mol, Purity >99%. Selective α₂A adrenoceptor antagonist (Ki values are 5.68 and 651 nM for α₂A and α₂B, respectively). Centrally active following subcutaneous administration. Demonstrates antidepressant and analgesic activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7246 Supplier Abcam Supplier No. AB12080610MG
MW 331.37 g/mol, Purity >99%. Selective α₂A adrenoceptor antagonist (Ki values are 5.68 and 651 nM for α₂A and α₂B, respectively). Centrally active following subcutaneous administration. Demonstrates antidepressant and analgesic activity.
The product is subject to the following: Abcam Restricted Use Statement