Learn More
Enzo Life Sciences BRL-50481 (10mg). CAS: 433695-36-4
SDP
Catalog No. 502011370
Encompass_Preferred
The first selective PDE7 inhibitor (Ki=180 nM). Formulation: Lyophilized. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-1370 Supplier Enzo Life Sciences Supplier No. BMLPD1200010
The first selective PDE7 inhibitor (Ki=180 nM). Formulation: Lyophilized. Purity: ≥98% (HPLC). Long Term Storage: Ambient.