Learn More
Medchemexpress LLC HY-109586 5mg Medchemexpress, BRL-50481 CAS:433695-36-4 Purity:>98%
SDP
Catalog No. 501968709
Encompass_Preferred
Medchemexpress, HY-109586 5mg BRL-50481 CAS:433695-36-4 BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8709 Supplier Medchemexpress LLC Supplier No. HY1095865MG
Medchemexpress, HY-109586 5mg BRL-50481 CAS:433695-36-4 BRL-50481 is a novel and selective inhibitor of PDE7 with IC50s of 0.15, 12.1, 62 and 490 μM for PDE7A, PDE7B, PDE4 and PDE3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.