Learn More
Medchemexpress LLC BRL 52537 hydrochloride | 112282-24-3 | 98.1% | 391.76 | C18H25Cl3N2O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10107925MG
Encompass_Preferred
BRL 52537 hydrochloride is a selective κ-opioid receptor (KOR) agonist supplied as the hydrochloride salt for research use. It is commonly used in pharmacology and neuroscience studies to probe KOR-mediated signaling and investigate neuroprotective effects; the compound is provided with reported high purity suitable for preclinical laboratory experiments.
  • Selective κ-opioid receptor agonist with high affinity.
  • Supplied as the hydrochloride salt for research use.
  • High reported purity suitable for laboratory experiments.
  • Available in small pre-weighed quantities for convenient dosing.
  • Used in pharmacology and neuroprotection research models.
Catalog No. 50-002-13032