Learn More
Medchemexpress LLC Brl 52537 hydrochloride | 112282-24-3 | MFCD00672679 | 98.1% | 391.76 | C18H25Cl3N2O | 5 MG
SDP
Catalog No. 5000213478
Encompass_Preferred
BRL 52537 hydrochloride is a potent, selective kappa-opioid receptor (KOR) agonist supplied as the hydrochloride salt for research use. It is provided as a solid with high reported purity and is intended for in vitro and in vivo pharmacology studies focusing on opioid receptor signaling and neuroprotection.
  • Selective kappa-opioid receptor agonist suitable for receptor pharmacology.
  • High reported purity of 98.1% for reliable experimental results.
  • Hydrochloride salt, solid form, stable for standard lab handling.
  • Available in small research quantities, e.g., 5 mg, for dose-ranging studies.
  • Molecular formula C18H25Cl3N2O and molecular weight 391.76 g/mol for stoichiometric calculations.
Catalog No. 50-002-13478 Supplier Medchemexpress LLC Supplier No. HY1010795MG
BRL 52537 hydrochloride is a potent, selective kappa-opioid receptor (KOR) agonist supplied as the hydrochloride salt for research use. It is provided as a solid with high reported purity and is intended for in vitro and in vivo pharmacology studies focusing on opioid receptor signaling and neuroprotection.
  • Selective kappa-opioid receptor agonist suitable for receptor pharmacology.
  • High reported purity of 98.1% for reliable experimental results.
  • Hydrochloride salt, solid form, stable for standard lab handling.
  • Available in small research quantities, e.g., 5 mg, for dose-ranging studies.
  • Molecular formula C18H25Cl3N2O and molecular weight 391.76 g/mol for stoichiometric calculations.