Learn More
Medchemexpress LLC BRL 52537 hydrochloride | 112282-24-3 | MFCD00672679 | 98.1% | 391.76 g/mol | C18H25Cl3N2O | 100 MG
SDP
Catalog No. 5000213466
Encompass_Preferred
BRL 52537 hydrochloride is a selective kappa-opioid receptor agonist research compound used to study opioid receptor pharmacology and neuroprotection. It reduces ischemia-evoked nitric oxide production and has demonstrated neuroprotective and antinociceptive effects in preclinical models, making it useful for both in vitro and in vivo neuroscience research.
  • Selective kappa-opioid receptor agonist with high affinity.
  • Reduces ischemia-evoked nitric oxide production in models.
  • Demonstrated neuroprotective and antinociceptive effects.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity for research applications.
Catalog No. 50-002-13466 Supplier Medchemexpress LLC Supplier No. HY101079100MG
BRL 52537 hydrochloride is a selective kappa-opioid receptor agonist research compound used to study opioid receptor pharmacology and neuroprotection. It reduces ischemia-evoked nitric oxide production and has demonstrated neuroprotective and antinociceptive effects in preclinical models, making it useful for both in vitro and in vivo neuroscience research.
  • Selective kappa-opioid receptor agonist with high affinity.
  • Reduces ischemia-evoked nitric oxide production in models.
  • Demonstrated neuroprotective and antinociceptive effects.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity for research applications.