Learn More
Abcam BRL 54443, agonist at 5-HT1E and 5-HT1F receptors, 10MG
Catalog No. 502707411
Encompass_Preferred
MW 230.31 Da, Purity >99%. Potent, high affinity and selective agonist at 5-HT1E/1F receptors (pKi values are 8.7 and 8.9, respectively). Displays >30-fold selectivity for human 5-HT over dopamine receptors (pKi values are 7.2 (5-HT1A), 6.9 (5-HT1B), 7.2 (5-HT1D), 5.9 (5-HT2A), 7.0 (5-HT2B), 6.5 (5-HT2C), 6.3 (D2) and 6.2 (D3)).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7411 Supplier Abcam Supplier No. AB12062710MG
MW 230.31 Da, Purity >99%. Potent, high affinity and selective agonist at 5-HT1E/1F receptors (pKi values are 8.7 and 8.9, respectively). Displays >30-fold selectivity for human 5-HT over dopamine receptors (pKi values are 7.2 (5-HT1A), 6.9 (5-HT1B), 7.2 (5-HT1D), 5.9 (5-HT2A), 7.0 (5-HT2B), 6.5 (5-HT2C), 6.3 (D2) and 6.2 (D3)).
The product is subject to the following: Abcam Restricted Use Statement