Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask Grad,1 L-AC Academic Line
SDP
Catalog No. NC3441069
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Narrow mouth with heavy duty rim to prevent chipping. Ideal for general laboratory use. graduated to approximate volumes.. MADE IN THE USA Size: 1l Stopper size: 9 Dimensions: 128x214mm
Catalog No. NC3441069 Supplier Bellco Glass, Inc. Supplier No. 250501000
Manufactured from type 1 class a Borosilicate glass. Narrow mouth with heavy duty rim to prevent chipping. Ideal for general laboratory use. graduated to approximate volumes.. MADE IN THE USA Size: 1l Stopper size: 9 Dimensions: 128x214mm