Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask Grad,500mL-AC Academic Line
SDP
Catalog No. 501945449
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Narrow mouth with heavy duty rim to prevent chipping. Ideal for general laboratory use. graduated to approximate volumes.. MADE IN THE USA Size: 500ml Stopper size: 7 Dimensions: 100x173mm
Catalog No. 50-194-5449 Supplier Bellco Glass, Inc. Supplier No. 250500500
Manufactured from type 1 class a Borosilicate glass. Narrow mouth with heavy duty rim to prevent chipping. Ideal for general laboratory use. graduated to approximate volumes.. MADE IN THE USA Size: 500ml Stopper size: 7 Dimensions: 100x173mm