Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,1000mL Narrow mouth
SDP
Catalog No. 501945442
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 1l Stopper size: 9. OD X OAH (MM):128 x 223. CASE QTY:6
Catalog No. 50-194-5442 Supplier Bellco Glass, Inc. Supplier No. 250401000
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 1l Stopper size: 9. OD X OAH (MM):128 x 223. CASE QTY:6