Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,2000mL Narrow mouth
SDP
Catalog No. 501945443
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 2l Stopper size: 10. OD X OAH (MM):158 x 290. CASE QTY:3
Catalog No. 50-194-5443 Supplier Bellco Glass, Inc. Supplier No. 250402000
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 2l Stopper size: 10. OD X OAH (MM):158 x 290. CASE QTY:3