Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,300mL Narrow mouth
SDP
Catalog No. 501945440
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 300ml Stopper size: 6. OD X OAH (MM):88 x 148. CASE QTY:12
Catalog No. 50-194-5440 Supplier Bellco Glass, Inc. Supplier No. 250400300
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Size: 300ml Stopper size: 6. OD X OAH (MM):88 x 148. CASE QTY:12