Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,50mL Narrow mouth
SDP
Catalog No. 501945437
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Sizes: 50ml Stopper size: 1. OD X OAH (MM):50 X 80. CASE QTY:24
Catalog No. 50-194-5437 Supplier Bellco Glass, Inc. Supplier No. 250400050
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes. MADE IN THE USA. Sizes: 50ml Stopper size: 1. OD X OAH (MM):50 X 80. CASE QTY:24