Learn More
Bellco Glass, Inc. Break Resistant Lip Erlenmeyer Flask-Grad,6000mL Narrow mouth
SDP
Catalog No. 501945446
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes.. MADE IN THE USA Size: 6l Stopper size: 10. OD X OAH (MM):237 x 395. CASE QTY:3
Catalog No. 50-194-5446 Supplier Bellco Glass, Inc. Supplier No. 250406000
Manufactured from type 1 class a Borosilicate glass. Ideal for general laboratory involving media mixing, storage and dispensing. Narrow mouth with break resistant lip that resists chipping. Graduated to approximate volumes.. MADE IN THE USA Size: 6l Stopper size: 10. OD X OAH (MM):237 x 395. CASE QTY:3