Learn More
Bellco Glass, Inc. Break Resistant Lip Media/Disp Flask WHC,2000mL Graduated
SDP
Catalog No. 501945506
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 2L Dimensions: 158x290mm
Catalog No. 50-194-5506 Supplier Bellco Glass, Inc. Supplier No. 253002000
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 2L Dimensions: 158x290mm