Learn More
Bellco Glass, Inc. Break Resistant Lip Media/Disp Flask WHC,250mL Graduated
SDP
Catalog No. 501945503
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 250mL Dimensions: 80x140mm
Catalog No. 50-194-5503 Supplier Bellco Glass, Inc. Supplier No. 253000250
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 250mL Dimensions: 80x140mm