Learn More
Bellco Glass, Inc. Break Resistant Lip Media/Disp Flask WHC,500mL Graduated
SDP
Catalog No. 501945504
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 500mL Dimensions: 100x183mm
Catalog No. 50-194-5504 Supplier Bellco Glass, Inc. Supplier No. 253000500
Manufactured from type 1 class a Borosilicate glass. Media storage and dispensing flask with bottom hose connection, accepts 1/2" tubing. Graduated to approximate volumes and the heavy duty break resistant neck provides an excellent fit for rubber stoppers. Not for vacuum use. . MADE IN THE USA Size: 500mL Dimensions: 100x183mm