Learn More
Bellco Glass, Inc. Break Resistant Lip Shake Flask W/3 Baffs,125mL
SDP
Catalog No. 501945518
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is designed for use with orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mlDimensions: 66x116mmCase Quantity: 12
Catalog No. 50-194-5518 Supplier Bellco Glass, Inc. Supplier No. 254100125
Manufactured from Type 1 Class A Borosilicate Glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is designed for use with orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mlDimensions: 66x116mmCase Quantity: 12