Learn More
Chemglass Life Sciences Flask, Shake, 1000mL, Baffled, Plain Top, Approx OD x Height (mm): 130 x 215
SDP
Catalog No. 501214590
Encompass_Preferred
Flask, Shake, 1000mL, Baffled, Plain Top, Approx OD x Height (mm): 130 x 215. Baffle indents on the lower outside edge of the flask to help oxygen transfer to the culture medium when used on an orbital shaker.
Catalog No. 50-121-4590 Supplier Chemglass Life Sciences Supplier No. CLS204005
Flask, Shake, 1000mL, Baffled, Plain Top, Approx OD x Height (mm): 130 x 215. Baffle indents on the lower outside edge of the flask to help oxygen transfer to the culture medium when used on an orbital shaker.