Learn More
Bellco Glass, Inc. Break Resistant Lip Shake Flask W/3 Baffs,2L
SDP
Catalog No. 501945521
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is designed for use with orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2LDimensions: 158x290mmCase Quantity:2541-02000: 3
Catalog No. 50-194-5521 Supplier Bellco Glass, Inc. Supplier No. 254102000
Manufactured from Type 1 Class A Borosilicate Glass with rigidly controlled baffle angles and depths. The 3 side baffles are used to promote optimal oxygen transfer while reducing vortex formation. Has a break resistant lip to resist chipping and is designed for use with orbital shakers.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2LDimensions: 158x290mmCase Quantity:2541-02000: 3