Learn More
Bellco Glass, Inc. Break Resistant Lip X-Dp BaffleFlask,300mL
SDP
Catalog No. 501945523
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 300mlDimensions: 88x150mmCase Quantity: 12Baffle Dimensions: 7mm X 35°
Catalog No. 50-194-5523 Supplier Bellco Glass, Inc. Supplier No. 254200300
Manufactured from Type 1 Class A Borosilicate Glass. Shake flasks are the basic tool for growth studies of aerobic microbial cultures. They have rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation. Designed for all orbital shakers. Break resistant lip to prevent chipping.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSize: 300mlDimensions: 88x150mmCase Quantity: 12Baffle Dimensions: 7mm X 35°