Learn More
Medchemexpress LLC Brm/Brg1 ATP inhibitor-1 | 2270879-17-7 | 99.7% | 10 MG
SDP
Catalog No. 5000233236
Encompass_Preferred
BRM/BRG1 ATP inhibitor-1 is an orally active, allosteric small-molecule inhibitor that selectively targets the ATPase activity of BRM (SMARCA2) and BRG1 (SMARCA4). It demonstrates sub-nanomolar potency and has shown anticancer activity in preclinical studies, making it useful for epigenetic and chromatin remodeling research.
  • Potent allosteric dual inhibitor of BRM and BRG1 ATPase activity.
  • Sub-nanomolar potency (IC50 < 0.005 μM) for both targets.
  • High purity suitable for biochemical and cell-based assays.
  • Available in solid and solution formats, including 10 mg solid and 10 mM in DMSO.
  • Recommended storage conditions preserve stability (powder: -20°C; in solvent: -80°C).
Catalog No. 50-002-33236 Supplier Medchemexpress LLC Supplier No. HY11937410MG
BRM/BRG1 ATP inhibitor-1 is an orally active, allosteric small-molecule inhibitor that selectively targets the ATPase activity of BRM (SMARCA2) and BRG1 (SMARCA4). It demonstrates sub-nanomolar potency and has shown anticancer activity in preclinical studies, making it useful for epigenetic and chromatin remodeling research.
  • Potent allosteric dual inhibitor of BRM and BRG1 ATPase activity.
  • Sub-nanomolar potency (IC50 < 0.005 μM) for both targets.
  • High purity suitable for biochemical and cell-based assays.
  • Available in solid and solution formats, including 10 mg solid and 10 mM in DMSO.
  • Recommended storage conditions preserve stability (powder: -20°C; in solvent: -80°C).