Learn More
Medchemexpress LLC HY-119374 5mg Medchemexpress, BRM/BRG1 ATP Inhibitor-1 CAS:2270879-17-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193745MG
Encompass_Preferred
Medchemexpress, HY-119374 5mg BRM/BRG1 ATP Inhibitor-1 CAS:2270879-17-7 BRM/BRG1 ATP Inhibitor-1 is an allosteric dual brahma homolog (BRM)/SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 2 (SMARCA2) and brahma related gene 1 (BRG1)/SMARCA4 ATPase activity inhibitor, both IC50s are below 0.005 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9533