Learn More
Selleck Chemical LLC BRM/BRG1 ATP Inhibitor-1
SDP
Catalog No. 503135586
Encompass_Preferred
BRM/BRG1 ATP Inhibitor-1 (Compound 14) is an orally active inhibitors of Brahma Homolog (BRM)/SMARCA2 (BRG1) with IC50s below 0 005 M
Catalog No. 50-313-5586 Supplier Selleck Chemical LLC Supplier No. E011110MM/1ML
BRM/BRG1 ATP Inhibitor-1 (Compound 14) is an orally active inhibitors of Brahma Homolog (BRM)/SMARCA2 (BRG1) with IC50s below 0 005 M