Learn More
Medchemexpress LLC BRM/BRG1 ATP Inhibitor-2 | 2368900-77-8 | 99.7% | C20H20N4O2S2 | 100 MG
SDP
Catalog No. 5000355742
Encompass_Preferred
BRM/BRG1 ATP Inhibitor-2 is a BRG1/BRM ATPase inhibitor for the research of BAF-related disorders. It is for research use only and not sold to patients.
  • BRG1/BRM ATPase inhibitor
  • For research of BAF-related disorders
  • Available as a white to off-white solid
  • Molecular weight is 412.53
  • For research use only
Catalog No. 50-003-55742 Supplier Medchemexpress LLC Supplier No. HY145946100MG
BRM/BRG1 ATP Inhibitor-2 is a BRG1/BRM ATPase inhibitor for the research of BAF-related disorders. It is for research use only and not sold to patients.
  • BRG1/BRM ATPase inhibitor
  • For research of BAF-related disorders
  • Available as a white to off-white solid
  • Molecular weight is 412.53
  • For research use only