Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Brm/Brg1 ATP inhibitor-2 | 2368900-77-8 | 99.7% | C20H20N4O2S2 | 10MG
SDP

Catalog No. 5000258674
Encompass_Preferred

BRM/BRG1 ATP Inhibitor-2 is a small-molecule inhibitor of the BRG1 and BRM ATPase subunits of the BAF (SWI/SNF) chromatin remodeling complex. It is supplied for research use to support studies of BAF-related disorders and investigations into epigenetic regulation and chromatin dynamics.

  • Small-molecule BRG1/BRM ATPase inhibitor for studies of chromatin remodeling.
  • 99.7% purity.
  • White to off-white solid appearance.
  • Molecular formula C20H20N4O2S2; molecular weight 412.53.
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or clinical use.
  • Available in multiple pack sizes suitable for screening and synthesis workflows.

Catalog No. 50-002-58674 Supplier Medchemexpress LLC Supplier No. HY14594610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BRM/BRG1 ATP Inhibitor-2 is a small-molecule inhibitor of the BRG1 and BRM ATPase subunits of the BAF (SWI/SNF) chromatin remodeling complex. It is supplied for research use to support studies of BAF-related disorders and investigations into epigenetic regulation and chromatin dynamics.

  • Small-molecule BRG1/BRM ATPase inhibitor for studies of chromatin remodeling.
  • 99.7% purity.
  • White to off-white solid appearance.
  • Molecular formula C20H20N4O2S2; molecular weight 412.53.
  • Storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use only; not for human or clinical use.
  • Available in multiple pack sizes suitable for screening and synthesis workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.