Learn More
Cayman Chemical BRMBRG1 ATP InhIbItr 1 500ug
SDP
Catalog No. 502911549
Encompass_Preferred
A dual inhibitor of BRM and BRG1 ATPase activities (IC50s = <0.005 µM for both); reduces expression of KRT80 in H1299 and RERF-LC-A1 cell lines; reduces tumor cell growth in a RERF-LC-A1 lung cancer mouse xenograft model at 20mg/kg
Catalog No. 50-291-1549 Supplier Cayman Chemical Supplier No. 36138500 UG
A dual inhibitor of BRM and BRG1 ATPase activities (IC50s = <0.005 µM for both); reduces expression of KRT80 in H1299 and RERF-LC-A1 cell lines; reduces tumor cell growth in a RERF-LC-A1 lung cancer mouse xenograft model at 20mg/kg