Learn More
Aviva Systems Biology Corp BRMS1 Antibody - C-terminal region (ARP63978_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63978_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRMS1. It was validated on Western Blot. . Alternate names: DKFZp564A063ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-288-135