Learn More
Aviva Systems Biology Corp BRMS1L Antibody - C-terminal region (ARP60404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRMS1L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BRMS1L. Alternate names: BRMS1, MGC11296ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-035