Learn More
Oxyrase Oxypras Brain Heart Infusion (Bhi) 50 X 5Mltubes 1/Pk
SDP
Catalog No. NC0982288
Encompass
Pre-reduced anaerobically sterilized nutritionally enriched medium. Utilized for growing anaerobes. Cell-free. Will not interfere with gram stain interpretation of cultures grown in it. Preferred broth medium for Oxyrase for Broth and AnaSelect Oxyrase for Broth.
Catalog No. NC0982288 Supplier Oxyrase Supplier No. BHIHK
Pre-reduced anaerobically sterilized nutritionally enriched medium. Utilized for growing anaerobes. Cell-free. Will not interfere with gram stain interpretation of cultures grown in it. Preferred broth medium for Oxyrase for Broth and AnaSelect Oxyrase for Broth.