Learn More
Med Vet International Broad Compression Plate 3.5, 10 holes, 122mm, J0422BE
Catalog No. 5000582872
Encompass_Preferred
3.5mm Broad Compression Plate. Uses the larger width 4.5mm compression plate with holes for 3.5mm cortical screws. A heavy-duty compression plate for bigger dogs.
Catalog No. 50-005-82872 Supplier Med Vet International Supplier No. JOR422BE
3.5mm Broad Compression Plate. Uses the larger width 4.5mm compression plate with holes for 3.5mm cortical screws. A heavy-duty compression plate for bigger dogs.