Learn More
Med Vet International Broad Compression Plate 3.5, 4 holes, 50mm, J0422C
Catalog No. 5000582954
Encompass_Preferred
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.
Catalog No. 50-005-82954 Supplier Med Vet International Supplier No. JOR422C
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.