Learn More
Med Vet International Broad Compression Plate 3.5, 5 holes, 62mm, J0422D
Catalog No. 5000582645
Encompass_Preferred
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.
Catalog No. 50-005-82645 Supplier Med Vet International Supplier No. JOR422D
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.