Learn More
Med Vet International Broad Plates 4.5mm, 16 hole, 263mm L, J0427NB
Catalog No. 5000582652
Encompass_Preferred
4.5mm Broad Plates. This is the strongest plate available forsmall animal orthopedics.
Catalog No. 50-005-82652 Supplier Med Vet International Supplier No. JOR427NB
4.5mm Broad Plates. This is the strongest plate available forsmall animal orthopedics.