Learn More
Med Vet International Broad Plates 4.5mm, 6 hole, 103mm L, J0427EB
Catalog No. 5000582971
Encompass_Preferred
4.5mm Broad Plates. This is the strongest plate available forsmall animal orthopedics.
Catalog No. 50-005-82971 Supplier Med Vet International Supplier No. JOR427EB
4.5mm Broad Plates. This is the strongest plate available forsmall animal orthopedics.