Learn More
Biotium AccuBlue Broad Range dsDNA Quantitation Kit
Catalog No. NC0956145
Encompass_Preferred
The AccuBlue Broad Range dsDNA Quantitation Kit provides reagents for 1500 microplate assays. This assay is intended for the quantification of dsDNA in the range of 2-2000 ng/uL.
Catalog No. NC0956145 Supplier Biotium Supplier No. 31007
The AccuBlue Broad Range dsDNA Quantitation Kit provides reagents for 1500 microplate assays. This assay is intended for the quantification of dsDNA in the range of 2-2000 ng/uL.