Learn More
Biotium AccuBlue Broad Range dsDNA Quantitation Solution
Supplier:  Biotium 31009
Encompass_Preferred
The AccuBlue Broad Range dsDNA Quantitation Kit provides reagents for 1500 microplate assays. This assay is intended for the quantification of dsDNA in the range of 2-2000 ng/uL.
Catalog No. NC0956147