Learn More
Biotium AccuBlue Broad Range dsDNA Quantitation Standards, set of nine, 0.5 mL each
Catalog No. NC0956149
Encompass_Preferred
Stand-alone set of the nine double-stranded DNA standards provided with the AccuBlue Broad Range dsDNA Quantitation Kit. Calf thymus dsDNA in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide at 0, 2, 6.25, 12.5, 25, 50, 100, 150, and 200 ng/uL, 0.5 mL each.
Catalog No. NC0956149 Supplier Biotium Supplier No. 31007C
Stand-alone set of the nine double-stranded DNA standards provided with the AccuBlue Broad Range dsDNA Quantitation Kit. Calf thymus dsDNA in 10 mM Tris pH 7.5, 1 mM EDTA, 2 mM sodium azide at 0, 2, 6.25, 12.5, 25, 50, 100, 150, and 200 ng/uL, 0.5 mL each.