Learn More
Proteintech Group Inc Broad range prestained protein marker
SDP
Supplier:  Proteintech Group Inc PL00002100UL
Encompass_Preferred
The PL00002 Broad Range Protein Marker is a ready-to-use three-color proteinstandard with 13 prestained proteins covering a wide range of molecular weightsfrom 3 to 245 kDa in Tris-Glycine buffer. The PL00002 Broad Range Prestained ProteinMarker is designed for monitoring protein separation during SDS-polyacrylamide gelelectrophoresis, verification of Western transfer efficiency on membranes(nitrocellulose, PVDF, or nylon) and for estimating the size of proteins.
Catalog No. 50-225-8722