Learn More
MSC P G Professional Sanitizer, EPA List-N, 3 pack, 1 gal
Catalog No. 502816097
Encompass
This all-purpose liquid sanitizer, with a sweet scent, is formulated to address fungus, germs, mildew, bacteria, and mold across various surfaces. It is packaged in 1-gallon bottles and is suitable for a wide range of material applications.
  • Formulated for broad-range sanitization against fungus, germs, mildew, bacteria, and mold
  • Suitable for vinyl, laminate, wood, fiberglass, enamel, glass, ceramic, plastic, and metal surfaces
  • Meets EPA List N and EPA Reg. No. 6836-278 standards
  • LEED applicable for sustainable practices
  • Supplied as a 3-pack of 1 gal bottles
Catalog No. 50-281-6097 Supplier MSC Supplier No. 10771459
This all-purpose liquid sanitizer, with a sweet scent, is formulated to address fungus, germs, mildew, bacteria, and mold across various surfaces. It is packaged in 1-gallon bottles and is suitable for a wide range of material applications.
  • Formulated for broad-range sanitization against fungus, germs, mildew, bacteria, and mold
  • Suitable for vinyl, laminate, wood, fiberglass, enamel, glass, ceramic, plastic, and metal surfaces
  • Meets EPA List N and EPA Reg. No. 6836-278 standards
  • LEED applicable for sustainable practices
  • Supplied as a 3-pack of 1 gal bottles