Learn More
Diagnostic Biosystems BROAD SPECTRUM Broad Spectrum Negative Control
Supplier:  Diagnostic Biosystems K053100
Encompass_Preferred
Negative controls are used to determine the specificity of an antibody. No staining should be observed when the primary antibody is omitted. This product is specifically designed to work with rabbit, mouse and goat antibodies. The Broad Spectrum Negative Control has been titered to work with Polymer-based Secondary Systems in manual and automated IHC platforms.This product is specifically designed to work with rabbit, mouse and goat antibodies. The Broad Spectrum Negative Control has been titered to work with Polymer-based Secondary Systems in manual and automated IHC platforms.
Catalog No. 50-240-7449