Learn More
PLANT CELL TECHNOLOGY Broad-spectrum biocide for plant tissue culture, in-vitro laboratory use only, Plant Preservative Mixture™, Clear Liquid, Anti-Contamination, 250mL Bottle
Supplier:  PLANT CELL TECHNOLOGY PPM250
Encompass_Preferred
Plant Preservative Mixture™ (PPM™) is a liquid additive designed to reduce microbial contamination in plant tissue culture and micropropagation applications. This 250mL bottle provides ready-to-use solution that integrates easily into standard culture media. PPM™ contains broad-spectrum antimicrobial and antifungal components compatible with a wide variety of plant species, including ornamentals, woody plants, and edible crops. It supports aseptic culture and enhances explant survival rates in tissue culture applications. Packaged in a durable, clear 250mL bottle with secure cap, PPM™ ensures safe handling and storage. Ideal for research laboratories, commercial propagation, and specialized plant cell culture applications. Stable under recommended storage conditions.
Catalog No. NC3905164